6-Bromonicotinoyl chloride ≥97%

Supplier: Matrix Scientific

114503-1G
102894-178EA 958.46 USD
102894-178
6-Bromonicotinoyl chloride ≥97%
6-Bromonicotinoyl chloride

Hazard: Corrosive

Formula: C₆H₃BrClNO
MDL Number: MFCD18382461
CAS Number: 740841-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR