5-Chloronicotinoyl chloride ≥97%

Supplier: Matrix Scientific

114497-1G
102894-166EA 1423.08 USD
102894-166
5-Chloronicotinoyl chloride ≥97%
5-Chloronicotinoyl chloride

Hazard: Irritant

Formula: C₆H₃Cl₂NO
MDL Number: MFCD13173962
CAS Number: 85320-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR