2-Bromoisonicotinoyl chloride ≥97%

Supplier: Matrix Scientific

114492-1G
102894-156EA 813.85 USD
102894-156
2-Bromoisonicotinoyl chloride ≥97%
2-Bromoisonicotinoyl chloride

Hazard: Irritant

Formula: C₆H₃BrClNO
MDL Number: MFCD07367878
CAS Number: 887578-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR