4-Chloro-3-methoxybenzoyl chloride ≥97%
Supplier: Matrix Scientific
114475-1G
102894-120EA
835.38
USD
102894-120
4-Chloro-3-methoxybenzoyl chloride ≥97%
4-Chloro-3-methoxybenzoyl chloride
Hazard: Irritant
Formula:
C₈H₆Cl₂O₂ |
MDL Number:
MFCD18393573 CAS Number: 39887-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...