4-(Ethylthio)benzoyl chloride ≥97%

Supplier: Matrix Scientific

114468-1G
102894-106EA 813.85 USD
102894-106
4-(Ethylthio)benzoyl chloride ≥97%
4-(Ethylthio)benzoyl chloride

Hazard: Irritant

Formula: C₉H₉ClOS
MDL Number: MFCD28099726
CAS Number: 108249-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR