2-(2-Methylpropane-2-sulfonyl)acetic acid ≥97%

Supplier: Matrix Scientific

114464-1G
102894-098EA 1029.23 USD
102894-098
2-(2-Methylpropane-2-sulfonyl)acetic acid ≥97%
2-(2-Methylpropane-2-sulfonyl)acetic acid

Hazard: Irritant

Formula: C₆H₁₂O₄S
MDL Number: MFCD06208069
CAS Number: 36610-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR