(R)-N-Fmoc-azetidine-2-carboxylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
(R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)azetidine-2-carboxylic acid
114460-1G
102894-090EA
2192.31
USD
102894-090
(R)-N-Fmoc-azetidine-2-carboxylic acid ≥97%
(R)-N-Fmoc-azetidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₉H₁₇NO₄ MW: 323.35 g/mol |
MDL Number:
MFCD01320875 CAS Number: 374791-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...