3-Cyclopentene-1,1-dicarboxylic acid ≥97%

Supplier: Matrix Scientific

114454-1G
102894-074EA 813.85 USD
102894-074
3-Cyclopentene-1,1-dicarboxylic acid ≥97%
3-Cyclopentene-1,1-dicarboxylic acid

Hazard: Irritant

Formula: C₇H₈O₄
MDL Number: MFCD20638529
CAS Number: 88326-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR