4-(Difluoromethyl)benzoic acid ≥97%
Supplier: Matrix Scientific
114449-1G
102894-062EA
400
USD
102894-062
4-(Difluoromethyl)benzoic acid ≥97%
4-(Difluoromethyl)benzoic acid
Hazard: Irritant
Formula:
C₈H₆F₂O₂ MW: 172.1289 g/mol |
MDL Number:
MFCD07784270 CAS Number: 55805-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...