4-Hydroxy-3,5-diiodobenzoic acid ≥97%

Supplier: Matrix Scientific

114439-10G
102894-040EA 303.08 USD
102894-040
4-Hydroxy-3,5-diiodobenzoic acid ≥97%
4-Hydroxy-3,5-diiodobenzoic acid

Hazard: Irritant

Formula: C₇H₄I₂O₃
MDL Number: MFCD00016532
CAS Number: 618-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR