3-(2-Aminopyrimidin-4-yl)benzoic acid ≥97%

Supplier: Matrix Scientific

114438-1G
102894-036EA 958.46 USD
102894-036
3-(2-Aminopyrimidin-4-yl)benzoic acid ≥97%
3-(2-Aminopyrimidin-4-yl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₉N₃O₂
MDL Number: MFCD09859835
CAS Number: 942035-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR