4-Phenylsulfanylbutyric acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Phenylthio)butanoic acid

114435-1G
102894-030EA 426.15 USD
102894-030
4-Phenylsulfanylbutyric acid ≥97%
4-Phenylsulfanylbutyric acid

Hazard: Irritant

Formula: C₁₀H₁₂O₂S
MW: 196.27 g/mol
MDL Number: MFCD02082831
CAS Number: 17742-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR