2-(1,1-Difluoroethyl)benzoic acid ≥97%

Supplier: Matrix Scientific

114430-1G
102894-020EA 1707.69 USD
102894-020
2-(1,1-Difluoroethyl)benzoic acid ≥97%
2-(1,1-Difluoroethyl)benzoic acid

Hazard: Irritant

Formula: C₉H₈F₂O₂
MDL Number: MFCD20659035
CAS Number: 892390-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR