4-(5-Oxazolyl)benzylamine ≥97%

Supplier: Matrix Scientific

114415-1G
102893-988EA 1443.08 USD
102893-988
4-(5-Oxazolyl)benzylamine ≥97%
4-(5-Oxazolyl)benzylamine

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD06213310
CAS Number: 672324-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR