6-Chloroimidazo[1,2-b]pyridazine-3-carbaldehyde ≥97%

Supplier: Matrix Scientific

114407-1G
102893-972EA 738.46 USD
102893-972
6-Chloroimidazo[1,2-b]pyridazine-3-carbaldehyde ≥97%
6-Chloroimidazo[1,2-b]pyridazine-3-carbaldehyde

Hazard: Irritant

Formula: C₇H₄ClN₃O
MDL Number: MFCD11044762
CAS Number: 154578-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR