4-Methoxy-2,5-dimethylbenzaldehyde ≥97%

Supplier: Matrix Scientific

114398-1G
102893-954EA 303.08 USD
102893-954
4-Methoxy-2,5-dimethylbenzaldehyde ≥97%
4-Methoxy-2,5-dimethylbenzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MDL Number: MFCD00003343
CAS Number: 6745-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR