2-(Dimethylaminomethyl)-3-hydroxypyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-((Dimethylamino)methyl)pyridin-3-ol

114397-10G
102893-952EA 118.46 USD
102893-952
2-(Dimethylaminomethyl)-3-hydroxypyridine ≥97%
2-(Dimethylaminomethyl)-3-hydroxypyridine

Hazard: Irritant

Formula: C₈H₁₂N₂O
MW: 152.2 g/mol
MDL Number: MFCD00006347
CAS Number: 2168-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR