N-Methyl-1-[6-(trifluoromethyl)pyridin-3-yl]methanamine ≥97%

Supplier: Matrix Scientific

Synonyms: N-Methyl-1-(6-(trifluoromethyl)pyridin-3-yl)methanamine

114396-1G
102893-948EA 1298.46 USD
102893-948
N-Methyl-1-[6-(trifluoromethyl)pyridin-3-yl]methanamine ≥97%
N-Methyl-1-[6-(trifluoromethyl)pyridin-3-yl]methanamine

Hazard: Irritant

Formula: C₈H₉F₃N₂
MW: 190.17 g/mol
MDL Number: MFCD07375086
CAS Number: 886364-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR