2-Chloro-N-pyridin-2-ylmethylacetamide ≥97%

Supplier: Matrix Scientific

114395-1G
102893-946EA 473.85 USD
102893-946
2-Chloro-N-pyridin-2-ylmethylacetamide ≥97%
2-Chloro-N-pyridin-2-ylmethylacetamide

Hazard: Irritant

Formula: C₈H₉ClN₂O
MDL Number: MFCD03381856
CAS Number: 46120-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR