Ethyl(pyridin-2-ylmethyl)amine ≥97%

Supplier: Matrix Scientific

Synonyms: N-(Pyridin-2-ylmethyl)ethanamine

114393-1G 114393-10G
102893-940EA 364.62 USD
102893-940 102893-942
Ethyl(pyridin-2-ylmethyl)amine ≥97%
Ethyl(pyridin-2-ylmethyl)amine

Hazard: Irritant

Formula: C₈H₁₂N₂
MW: 136.2 g/mol
MDL Number: MFCD07411568
CAS Number: 51639-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR