N-[4-(4-Methylpiperazin-1-yl)cyclohexyl]acetamide ≥97%

Supplier: Matrix Scientific

114385-1G
102893-920EA 958.46 USD
102893-920
N-[4-(4-Methylpiperazin-1-yl)cyclohexyl]acetamide ≥97%
N-[4-(4-Methylpiperazin-1-yl)cyclohexyl]acetamide

Hazard: Irritant

Formula: C₁₃H₂₅N₃O
MDL Number: MFCD28099721
CAS Number: 1214265-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR