2-Amino-1-pyrrolidine hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3,4-Dihydro-2H-pyrrol-5-amine hydrochloride2-Amino-1-pyrroline hydrochloride

114380-1G
102893-910EA 523.08 USD
102893-910
2-Amino-1-pyrrolidine hydrochloride ≥97%
2-Amino-1-pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₄H₉ClN₂
MW: 120.58 g/mol
MDL Number: MFCD00790772
CAS Number: 7544-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR