4-Amino-2-chloro-5-methylpyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-5-methylpyridin-4-amine
114371-1G
102893-892EA
1443.08
USD
102893-892
4-Amino-2-chloro-5-methylpyridine ≥97%
4-Amino-2-chloro-5-methylpyridine
Hazard: Irritant
Formula:
C₆H₇ClN₂ MW: 142.5862 g/mol |
MDL Number:
MFCD11977435 CAS Number: 79055-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...