2,6-Dichloro-4-phenylaniline ≥97%

Supplier: Matrix Scientific

114368-1G
102893-884EA 813.85 USD
102893-884
2,6-Dichloro-4-phenylaniline ≥97%
2,6-Dichloro-4-phenylaniline

Hazard: Irritant

Formula: C₁₂H₉Cl₂N
MDL Number: MFCD09042411
CAS Number: 134006-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR