2,3,4,5-Tetrachloroaniline ≥97%

Supplier: Matrix Scientific

114365-1G
102893-878EA 473.85 USD
102893-878
2,3,4,5-Tetrachloroaniline ≥97%
2,3,4,5-Tetrachloroaniline

Hazard: Irritant

Formula: C₆H₃Cl₄N
MW: 230.91 g/mol
CAS Number: 634-83-3
UN: 1648
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR