4-(2,2,2-Trifluoroethyl)aniline ≥97%

Supplier: Matrix Scientific

114364-1G
102893-876EA 1069.23 USD
102893-876
4-(2,2,2-Trifluoroethyl)aniline ≥97%
4-(2,2,2-Trifluoroethyl)aniline

Hazard: Irritant

Formula: C₈H₈F₃N
MDL Number: MFCD04972985
CAS Number: 131395-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR