2-Bromo-4-(pentafluorothio)aniline ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Amino-3-bromophenylsulphur pentafluoride

114357-1G
102893-862EA 690.77 USD
102893-862
2-Bromo-4-(pentafluorothio)aniline ≥97%
2-Bromo-4-(pentafluorothio)aniline

Hazard: Irritant

Formula: C₆H₅BrF₅NS
MDL Number: MFCD20731078
CAS Number: 159727-25-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR