4-(Isopropylsulfonyl)aniline ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Propane-2-sulfonyl)aniline4-(Propane-2-sulphonyl)aniline

114356-1G
102893-860EA 1298.46 USD
102893-860
4-(Isopropylsulfonyl)aniline ≥97%
4-(Isopropylsulphonyl)aniline

Hazard: Irritant

Formula: C₉H₁₃NO₂S
MDL Number: MFCD01566617
CAS Number: 25355-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR