2-Bromo-1-(4-(phenylthio)phenyl)ethanone ≥97%

Supplier: Matrix Scientific

114355-1G
102893-858EA 884.62 USD
102893-858
2-Bromo-1-(4-(phenylthio)phenyl)ethanone ≥97%
2-Bromo-1-(4-(phenylthio)phenyl)ethanone

Hazard: Irritant

Formula: C₁₄H₁₁BrOS
MDL Number: MFCD08282296
CAS Number: 28179-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR