2-(3-Nitrophenyl)propanoyl chloride ≥97%

Supplier: Matrix Scientific

114352-1G
102893-852EA 1589.23 USD
102893-852
2-(3-Nitrophenyl)propanoyl chloride ≥97%
2-(3-Nitrophenyl)propanoyl chloride

Hazard: Irritant

Formula: C₉H₈ClNO₃
MDL Number: MFCD23096909
CAS Number: 156516-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR