2-Hydroxybenzoyl chloride ≥97%

Supplier: Matrix Scientific

114351-1G
102893-850EA 1369.23 USD
102893-850
2-Hydroxybenzoyl chloride ≥97%
2-Hydroxybenzoyl chloride

Hazard: Irritant/Stored under Argon

Formula: C₇H₅ClO₂
MDL Number: MFCD11169645
CAS Number: 1441-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR