1-(3-Methoxyphenyl)cyclopropanecarbonyl chloride ≥97%

Supplier: Matrix Scientific

114342-1G
102893-832EA 1395.38 USD
102893-832
1-(3-Methoxyphenyl)cyclopropanecarbonyl chloride ≥97%
1-(3-Methoxyphenyl)cyclopropanecarbonyl chloride

Hazard: Irritant

Formula: C₁₁H₁₁ClO₂
MDL Number: MFCD18907433
CAS Number: 63201-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR