4-Phenylsulfanylbutyroyl chloride ≥97%

Supplier: Matrix Scientific

114337-1G
102893-822EA 958.46 USD
102893-822
4-Phenylsulfanylbutyroyl chloride ≥97%
4-Phenylsulfanylbutyroyl chloride

Hazard: Irritant

Formula: C₁₀H₁₁ClOS
MDL Number: MFCD11939630
CAS Number: 138738-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR