2-Methyl-5-nitrobenzoyl chloride ≥97%

Supplier: Matrix Scientific

114332-1G
102893-810EA 738.46 USD
102893-810
2-Methyl-5-nitrobenzoyl chloride ≥97%
2-Methyl-5-nitrobenzoyl chloride

Hazard: Irritant

Formula: C₈H₆ClNO₃
MDL Number: MFCD03701211
CAS Number: 64688-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR