2-Fluoro-6-hydroxybenzoyl chloride ≥97%

Supplier: Matrix Scientific

114331-1G
102893-808EA 813.85 USD
102893-808
2-Fluoro-6-hydroxybenzoyl chloride ≥97%
2-Fluoro-6-hydroxybenzoyl chloride

Hazard: Irritant

Formula: C₇H₄ClFO₂
MDL Number: MFCD13173862
CAS Number: 214752-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR