5-Bromo-2-fluorophenylacetyl chloride ≥97%

Supplier: Matrix Scientific

114329-1G
102893-804EA 958.46 USD
102893-804
5-Bromo-2-fluorophenylacetyl chloride ≥97%
5-Bromo-2-fluorophenylacetyl chloride

Hazard: Irritant

Formula: C₈H₅BrClFO
MDL Number: MFCD26686419
CAS Number: 1346146-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR