2-Chloro-6-fluorophenylacetyl chloride ≥97%

Supplier: Matrix Scientific

114327-1G
102893-800EA 570.77 USD
102893-800
2-Chloro-6-fluorophenylacetyl chloride ≥97%
2-Chloro-6-fluorophenylacetyl chloride

Hazard: Corrosive/Light Sensitive/Moisture Sensitive

Formula: C₈H₅Cl₂FO
MW: 207.03 g/mol
MDL Number: MFCD11188513
CAS Number: 179314-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR