2-Methyl-2-phenylpropanoyl chloride ≥97%
Supplier: Matrix Scientific
114319-1G
102893-782EA
570.77
USD
102893-782
2-Methyl-2-phenylpropanoyl chloride ≥97%
2-Methyl-2-phenylpropanoyl chloride
Hazard: Corrosive
Formula:
C₁₀H₁₁ClO |
MDL Number:
MFCD09903682 CAS Number: 36293-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...