[1,1'-Biphenyl]-2,2'-dicarbonyl dichloride ≥97%

Supplier: Matrix Scientific

114311-1G
102898-404EA 813.85 USD
102898-404
[1,1'-Biphenyl]-2,2'-dicarbonyl dichloride ≥97%
[1,1'-Biphenyl]-2,2'-dicarbonyl dichloride

Hazard: Irritant

Formula: C₁₄H₈Cl₂O₂
MDL Number: MFCD11617025
CAS Number: 7535-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR