3-Bromo-5-fluorobenzoyl chloride ≥97%

Supplier: Matrix Scientific

114285-1G
102893-738EA 253.85 USD
102893-738
3-Bromo-5-fluorobenzoyl chloride ≥97%
3-Bromo-5-fluorobenzoyl chloride

Hazard: Irritant

Formula: C₇H₃BrClFO
MDL Number: MFCD07368699
CAS Number: 887266-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR