3-Hydroxy-4-methylbenzoyl chloride ≥97%

Supplier: Matrix Scientific

114282-1G
102893-732EA 958.46 USD
102893-732
3-Hydroxy-4-methylbenzoyl chloride ≥97%
3-Hydroxy-4-methylbenzoyl chloride

Hazard: Irritant

Formula: C₈H₇ClO₂
MDL Number: MFCD11169649
CAS Number: 348165-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR