N-(2,3-Dihydroxypropyl)-2,2,2-trifluoroacetamide ≥97%

Supplier: Matrix Scientific

114276-1G
102893-718EA 958.46 USD
102893-718
N-(2,3-Dihydroxypropyl)-2,2,2-trifluoroacetamide ≥97%
N-(2,3-Dihydroxypropyl)-2,2,2-trifluoroacetamide

Hazard: Irritant

Formula: C₅H₈F₃NO₃
MDL Number: MFCD21709293
CAS Number: 125348-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR