2-Azabicyclo[2.2.1]heptane ≥97%
Supplier: Matrix Scientific
114264-1G
102893-684EA
1175.38
USD
102893-684
2-Azabicyclo[2.2.1]heptane ≥97%
2-Azabicyclo[2.2.1]heptane
Hazard: Irritant
Formula:
C₆H₁₁N |
MDL Number:
MFCD05864542 CAS Number: 279-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...