Isoindoline ≥97%

Supplier: Matrix Scientific

114259-1G
102893-672EA 329.23 USD
102893-672
Isoindoline ≥97%
Isoindoline

Hazard: Irritant

Formula: C₈H₉N
Boiling Pt: 126…127 °C (18 Torr)
Melting Pt: 17 °C
Density: 1.05
MDL Number: MFCD00605324
CAS Number: 496-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR