3-Azido-7-hydroxycoumarin ≥97%

Supplier: Matrix Scientific

114258-1G
102893-670EA 2596.92 USD
102893-670
3-Azido-7-hydroxycoumarin ≥97%
3-Azido-7-hydroxycoumarin

Hazard: Irritant

Formula: C₉H₅N₃O₃
Melting Pt: 118…120 °C
MDL Number: MFCD12924840
CAS Number: 817638-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR