3-Amino-2-phenylindenone ≥97%

Supplier: Matrix Scientific

114253-1G
102893-658EA 86.15 USD
102893-658
3-Amino-2-phenylindenone ≥97%
3-Amino-2-phenylindenone

Hazard: Irritant

Formula: C₁₅H₁₁NO
MDL Number: MFCD00051614
CAS Number: 1947-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR