7,9-Dihydro-8H-purin-8-one ≥97%

Supplier: Matrix Scientific

114252-1G
102893-656EA 1153.85 USD
102893-656
7,9-Dihydro-8H-purin-8-one ≥97%
7,9-Dihydro-8H-purin-8-one

Hazard: Irritant

Formula: C₅H₄N₄O
MDL Number: MFCD18823025
CAS Number: 13230-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR