2-Methylene-1,3-dioxepane ≥97%

Supplier: Matrix Scientific

114246-1G
102893-644EA 1298.46 USD
102893-644
2-Methylene-1,3-dioxepane ≥97%
2-Methylene-1,3-dioxepane

Hazard: Irritant

Formula: C₆H₁₀O₂
MDL Number: MFCD00467150
CAS Number: 69814-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR