4-Acetyl-1H-imidazole ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Acetylimidazole

114238-1G
102893-630EA 150.77 USD
102893-630
4-Acetyl-1H-imidazole ≥97%
4-Acetyl-1H-imidazole

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Melting Pt: 169…170 °C
Density: 1.19
MDL Number: MFCD09033077
CAS Number: 61985-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR