1-(1-Methoxypropan-2-yl)-1H-pyrrole ≥97%

Supplier: Matrix Scientific

114228-1G
102893-610EA 690.77 USD
102893-610
1-(1-Methoxypropan-2-yl)-1H-pyrrole ≥97%
1-(1-Methoxypropan-2-yl)-1H-pyrrole

Hazard: Irritant

Formula: C₈H₁₃NO
MDL Number: MFCD06334613
CAS Number: 745026-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR